missing translation for 'onlineSavingsMsg'
Learn More

IRAK inhibitor 2, MedChemExpress

Product Code. 16468307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16468307 5 mg 5mg
16415618 10 mg 10mg
16401787 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 16468307

Brand: Medchem Express HY13276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IRAK inhibitor 2 is interleukin-1 receptor associated kinase inhibitor .

IRAK inhibitor 2 is interleukin-1 receptor associated kinase inhibitor .

Specifications

Chemical Name or Material IRAK inhibitor 2
CAS 928333-30-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H14N4O2
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (163.23 mM)
SMILES OC1=CC=C(C=C1)C2=CN=C3C=CC(NCC4=CC=CO4)=NN32
Molecular Weight (g/mol) 306.32
Formula Weight 306.32
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.