missing translation for 'onlineSavingsMsg'
Learn More

IMD-0354, MedChemExpress

Product Code. 16406257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16406257 5 mg 5mg
16463528 10 mg 10mg
16439966 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16406257 Supplier Medchem Express Supplier No. HY10172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IMD-0354 (IKK2 Inhibitor V) is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC50 of 1.2 uM.

IMD-0354 (IKK2 Inhibitor V) is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC50 of 1.2 uM.

Specifications

Chemical Name or Material IMD-0354
CAS 978-62-1
Color White
Purity Grade Notes Research
Molecular Formula C15H8ClF6NO2
Quantity 5 mg
Synonym IKK2 Inhibitor V
Solubility Information DMSO : ≥ 100 mg/mL (260.64 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1)C2=CC(Cl)=CC=C2O
Molecular Weight (g/mol) 383.67
Formula Weight 383.67
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.