missing translation for 'onlineSavingsMsg'
Learn More

GAL-021, MedChemExpress

Product Code. 16416087
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416087

Brand: Medchem Express HY101422

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GAL-021 is a potent BKCa-channel blocker. GAL-021 inhibits KCa1.1 in GH3 cells. GAL-021 is a novel breathing control modulator that is based on selective modification of the almitrine pharmacophore. GAL-021 increases minute ventilation in rats and non-human primates.

GAL-021 is a potent BKCa-channel blocker. GAL-021 inhibits KCa1.1 in GH3 cells. GAL-021 is a novel breathing control modulator that is based on selective modification of the almitrine pharmacophore. GAL-021 increases minute ventilation in rats and non-human primates.

Specifications

Chemical Name or Material GAL-021
Physical Form Solid
CAS 1380341-99-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H22N6O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 30 mg/mL (117.96 mM)
SMILES CCCNC1=NC(N(OC)C)=NC(NCCC)=N1
Molecular Weight (g/mol) 254.33
Formula Weight 254.33
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.