missing translation for 'onlineSavingsMsg'
Learn More

DDR1-IN-1, MedChemExpress

Product Code. 16428577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16428577

Brand: Medchem Express HY139795mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DDR1-IN-1 is a potent and selective DDR1 receptor tyrosine kinase inhibitor with an IC50 of 105 nM; 4-fold less potent for DDR2 (IC50 = 413 nM).

DDR1-IN-1 is a potent and selective DDR1 receptor tyrosine kinase inhibitor with an IC50 of 105 nM

Specifications

Chemical Name or Material DDR1-IN-1
CAS 1449685-96-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H31F3N4O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (180.97 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C)C(OC2=CC3=C(NC(C3)=O)C=C2)=C1)C4=CC=C(CN5CCN(CC)CC5)C(C(F)(F)F)=C4
Molecular Weight (g/mol) 552.59
Formula Weight 552.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.