missing translation for 'onlineSavingsMsg'
Learn More

BAM 15, MedChemExpress

Product Code. 16457097
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. Quantity unitSize
16457097 5 mg 5mg
16443056 25 mg 25mg
16404378 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16457097

Brand: Medchem Express HY110284

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAM 15 is a mitochondrial protonophore uncoupler. BAM 15 is an oxidative phosphorylation (OXPHOS) uncoupler.

BAM 15 is a mitochondrial protonophore uncoupler. BAM 15 is an oxidative phosphorylation (OXPHOS) uncoupler.

Specifications

Chemical Name or Material BAM 15
CAS 210302-17-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H10F2N6O
Quantity 5 mg
Solubility Information DMSO : 75 mg/mL (220.40 mM; Need ultrasonic)
SMILES FC(C=CC=C1)=C1NC2=NC3=NON=C3N=C2NC4=C(C=CC=C4)F
Molecular Weight (g/mol) 340.29
Formula Weight 340.29
Percent Purity 99.26%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.