Learn More
Nudifloramide, MedChemExpress
Product Code. 16457517
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity unitSize
16457517 5 mg 5mg
1 options
Product Code. 16457517
Brand: Medchem Express HY113432
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Nudifloramide (2PY) is one of the end products of nicotinamide-adenine dinucleotide (NAD) degradation. Nudifloramide significantly inhibits poly(ADP-ribose) polymerase (PARP-1) activity in vitro.
Nudifloramide (2PY) is one of the end products of nicotinamide-adenine dinucleotide (NAD) degradation. Nudifloramide significantly inhibits poly(ADP-ribose) polymerase (PARP-1) activity in vitro.
Specifications
Chemical Name or Material Nudifloramide
Physical Form Solid
CAS 701-44-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C7H8N2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym 2PY
Solubility Information DMSO : 62.5 mg/mL (410.78 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (219.06 mM; Need ultrasonic)
SMILES O=C(C(C=C1)=CN(C)C1=O)N
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Percent Purity 98.13%
Grade Research