missing translation for 'onlineSavingsMsg'
Learn More

Itopride hydrochloride, MedChemExpress

Product Code. 16441048
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 16441048

Brand: Medchem Express HYB0732

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Itopride (HSR803) hydrochloride is a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. Itopride hydrochloride enhances gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions, can be used as a gastrointestinal prokinetic agent. Itopride can be used for researching gastro-esophageal reflux disease (GERD).

Itopride (HSR803) hydrochloride is a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. Itopride hydrochloride enhances gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions, can be used as a gastrointestinal prokinetic agent. Itopride can be used for researching gastro-esophageal reflux disease (GERD).

Specifications

Chemical Name or Material Itopride hydrochloride
CAS 122892-31-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H27ClN2O4
Quantity 5 g
Synonym HSR803
Solubility Information DMSO : 100 mg/mL (253.24 mM; Need ultrasonic) ∣H2O : 50 mg/mL (126.62 mM; Need ultrasonic)
SMILES O=C(NCC1=CC=C(OCCN(C)C)C=C1)C2=CC=C(OC)C(OC)=C2.Cl
Molecular Weight (g/mol) 394.89
Formula Weight 394.89
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.