missing translation for 'onlineSavingsMsg'
Learn More

DL-Lysine, MedChemExpress

Product Code. 16441158
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441158 5 g 5g
16432399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441158 Supplier Medchem Express Supplier No. HYB2236

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress DL-Lysine is a racemic mixture of the D-Lysine and L-Lysine. Lysine is an α-amino acid that is used in the biosynthesis of proteins.

DL-Lysine is a racemic mixture of the D-Lysine and L-Lysine. Lysine is an α-amino acid that is used in the biosynthesis of proteins.

Specifications

Chemical Name or Material DL-Lysine
CAS 70-54-2
Color White
Purity Grade Notes Research
Molecular Formula C6H14N2O2
Synonym (±)-2,6-Diaminocaproic acid
Solubility Information H2O : 100 mg/mL (684.04 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (6.84 mM; ultrasonic and warming and heat to 80°C)
SMILES NC(CCCCN)C(O)=O
Molecular Weight (g/mol) 146.19
Quantity 5 g
Formula Weight 146.19
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.