missing translation for 'onlineSavingsMsg'
Learn More

TAK-901, MedChemExpress

Product Code. 16431257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431257 50 mg 50mg
16487737 5 mg 5mg
16465766 100 mg 100mg
16435018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16431257 Supplier Medchem Express Supplier No. HY12201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TAK-901 is a multi-targeted aurora inhibitor with IC50s of 21 and 15 nM for aurora A and B, respectively.

TAK-901 is a multi-targeted aurora inhibitor with IC50s of 21 and 15 nM for aurora A and B, respectively.

Specifications

Chemical Name or Material TAK-901
CAS 934541-31-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H32N4O3S
Quantity 50 mg
Solubility Information DMSO : 2 mg/mL (3.96 mM; Need ultrasonic and warming)
SMILES O=C(NC1CCN(CC1)C)C2=C(C3=C(C4=CC(C)=CN=C4N3)C(C5=CC=CC(S(=O)(CC)=O)=C5)=C2)C
Molecular Weight (g/mol) 504.64
Formula Weight 504.64
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.