missing translation for 'onlineSavingsMsg'
Learn More

T0901317, MedChemExpress

Product Code. 16400437
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16400437

Brand: Medchem Express HY10626

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress T0901317 is an orally active and highly selective LXR agonist with an EC50 of 20 nM for LXRα. T0901317 activates FXR with an EC50 of 5 μM. T0901317 is RORα and RORγ dual inverse agonist with Ki values of 132 nM and 51 nM, respectively. T0901317 induces apoptosis and inhibits the development of atherosclerosis in low-density lipoprotein (LDL) receptor-deficient mice.

T0901317 is an orally active and highly selective LXR agonist with an EC50 of 20 nM for LXRα. T0901317 activates FXR with an EC50 of 5 μM. T0901317 is RORα and RORγ dual inverse agonist with Ki values of 132 nM and 51 nM, respectively. T0901317 induces apoptosis and inhibits the development of atherosclerosis in low-density lipoprotein (LDL) receptor-deficient mice.

Specifications

Chemical Name or Material T0901317
CAS 293754-55-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H12F9NO3S
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (207.76 mM; Need ultrasonic)
SMILES OC(C(F)(F)F)(C(F)(F)F)C1=CC=C(N(CC(F)(F)F)S(C2=CC=CC=C2)(=O)=O)C=C1
Molecular Weight (g/mol) 481.33
Formula Weight 481.33
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.