missing translation for 'onlineSavingsMsg'
Learn More

SNS-314 mesylate, MedChemExpress

Product Code. 16431157
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16431157 50 mg 50mg
16467657 5 mg 5mg
16434908 10 mg 10mg
16435686 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16431157

Brand: Medchem Express HY12003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SNS-314 mesylate is a potent and selective aurora kinase inhibitor with IC50s of 9, 31, and 6 nM for aurora A, B and C, respectively.

SNS-314 mesylate is a potent and selective aurora kinase inhibitor with IC50s of 9, 31, and 6 nM for aurora A, B and C, respectively.

Specifications

Chemical Name or Material SNS-314 mesylate
CAS 1146618-41-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H19ClN6O4S3
Quantity 50 mg
Solubility Information DMSO : 150 mg/mL (284.61 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC(Cl)=C1)NC2=NC=C(S2)CCNC3=C4C(C=CS4)=NC=N3.OS(C)(=O)=O
Molecular Weight (g/mol) 527.04
Formula Weight 527.04
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.