missing translation for 'onlineSavingsMsg'
Learn More

Adezmapimod hydrochloride, MedChemExpress

Product Code. 16450127
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16450127

Brand: Medchem Express HY10256A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Adezmapimod (SB 203580) hydrochloride is a selective and ATP-competitive p38 MAPK inhibitor with IC50s of 50 nM and 500 nM for SAPK2a/p38 and SAPK2b/p38β2, respectively. Adezmapimod hydrochloride inhibits LCK, GSK3β and PKBα with IC50s of 100-500-fold higher than that for SAPK2a/p38. Adezmapimod hydrochloride is an autophagy and mitophagy activator.

Adezmapimod (SB 203580) hydrochloride is a selective and ATP-competitive p38 MAPK inhibitor with IC50s of 50 nM and 500 nM for SAPK2a/p38 and SAPK2b/p38β2, respectively. Adezmapimod hydrochloride inhibits LCK, GSK3β and PKBα with IC50s of 100-500-fold higher than that for SAPK2a/p38. Adezmapimod hydrochloride is an autophagy and mitophagy activator.

Specifications

Chemical Name or Material Adezmapimod hydrochloride
CAS 869185-85-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H17ClFN3OS
Quantity 50 mg
Synonym SB 203580 hydrochlorideRWJ 64809 hydrochloride
Solubility Information DMSO : 100 mg/mL (241.60 mM; Need ultrasonic) ∣H2O : 8.43 mg/mL (20.37 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=C(C2=NC(C3=CC=C(F)C=C3)=C(C4=CC=NC=C4)N2)C=C1)C.[H]Cl
Molecular Weight (g/mol) 413.9
Formula Weight 413.9
Percent Purity 98.6%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.