missing translation for 'onlineSavingsMsg'
Learn More

Remetinostat, MedChemExpress

Product Code. 16449536
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449536 50 mg 50mg
16455707 5 mg 5mg
16434286 100 mg 100mg
16432968 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16449536 Supplier Medchem Express Supplier No. HY100365
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Remetinostat (SHP-141) is a hydroxamic acid-based inhibitor of histone deacetylase enzymes (HDAC) which is under development for the treatment of cutaneous T-cell lymphoma.

Remetinostat (SHP-141) is a hydroxamic acid-based inhibitor of histone deacetylase enzymes (HDAC) which is under development for the treatment of cutaneous T-cell lymphoma.

Specifications

Chemical Name or Material Remetinostat
CAS 946150-57-8
Color White
Purity Grade Notes Research
Molecular Formula C16H21NO6
Quantity 50 mg
Synonym SHP-141
Solubility Information DMSO : 150 mg/mL (463.91 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=C(OC(CCCCCCC(NO)=O)=O)C=C1
Molecular Weight (g/mol) 323.34
Formula Weight 323.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.