missing translation for 'onlineSavingsMsg'
Learn More

Potassium clavulanate cellulose, MedChemExpress

Product Code. 16423917
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16423917

Brand: Medchem Express HY19964

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.

Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, is a beta-lactamase inhibitor.

Specifications

Chemical Name or Material Potassium clavulanate cellulose
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H9NO5K.(C6H10O5)n
Quantity 50 mg
Synonym Potassium clavulanate:cellulose (1:1)
Solubility Information DMSO : 1 mg/mL (ultrasonic and warming and heat to 80°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O[C@H]1[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](C)[C@@H](CO)O2)[C@@H](CO)O[C@@H](OC)[C@@H]1O.O=C([C@@H](/C(O[C@]3([H])C4)=C/CO)N3C4=O)O[K].[n]
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.