missing translation for 'onlineSavingsMsg'
Learn More

ML240, MedChemExpress

Product Code. 16433857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433857 50 mg 50mg
16467908 10 mg 10mg
16410378 5 mg 5mg
16407726 100 mg 100mg
16403696 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16433857 Supplier Medchem Express Supplier No. HY19795

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML240 is a potent p97 inhibitor, inhibiting p97 ATPase with IC50 value of 100 nM.

ML240 is a potent p97 inhibitor, inhibiting p97 ATPase with IC50 value of 100 nM.

Specifications

Chemical Name or Material ML240
CAS 1346527-98-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H20N6O
Quantity 50 mg
Solubility Information DMSO : 12.5 mg/mL (31.53 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble)
SMILES COC1=CC=CC2=C(NCC3=CC=CC=C3)N=C(N4C(N)=NC5=CC=CC=C45)N=C12
Molecular Weight (g/mol) 396.44
Formula Weight 396.44
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.