missing translation for 'onlineSavingsMsg'
Learn More

Ipidacrine Hydrochloride Hydrate, TRC

Product Code. 30009592
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009592 50 mg 50mg
30137146 1 g 1g
30004161 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009592 Supplier Toronto Research Chemicals Supplier No. TRCI73933350MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118499-70-0
Molecular Formula C12 H16 N2 . Cl H . H2 O
Molecular Weight (g/mol) 242.75
Synonym 1H-Cyclopenta[b]quinolin-9-amine, 2,3,5,6,7,8-hexahydro-, hydrochloride, hydrate (1:1:1), 1H-Cyclopenta[b]quinolin-9-amine, 2,3,5,6,7,8-hexahydro-, monohydrochloride, monohydrate (9CI), Ipidacrine hydrochloride hydrate
IUPAC Name 2,3,5,6,7,8-hexahydro-1H-cyclopenta[b]quinolin-9-amine;hydrate;hydrochloride
SMILES O.Cl.Nc1c2CCCc2nc3CCCCc13

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H16N2.ClH.H2O/c13-12-8-4-1-2-6-10(8)14-11-7-3-5-9(11)12;;/h1-7H2,(H2,13,14);1H;1H2
Formula Weight 242.1186
Percent Purity >95%
Chemical Name or Material Ipidacrine Hydrochloride Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.