Learn More
IMD-0560, MedChemExpress
Product Code. 16430407
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16430407
Brand: Medchem Express HY105661
MedChemExpress IMD-0560 is a novel IκB kinase β inhibitor.
IMD-0560 is a novel IκB kinase β inhibitor.
Specifications
Chemical Name or Material IMD-0560
CAS 439144-66-8
Color White
Purity Grade Notes Research
Molecular Formula C15H8BrF6NO2
Quantity 50 mg
Solubility Information Ethanol : 50 mg/mL (116.79 mM; Need ultrasonic)
SMILES O=C(C1=C(C=CC(Br)=C1)O)NC2=C(C=CC(C(F)(F)F)=C2)C(F)(F)F
Molecular Weight (g/mol) 428.12
Formula Weight 428.12
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.