missing translation for 'onlineSavingsMsg'
Learn More

GLPG0634 analog, MedChemExpress

Product Code. 16432087
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16432087

Brand: Medchem Express HY13961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GLPG0634 analog (Compoun 176) is a broad spectrum JAK inhibitor with IC50 values of <100 nM against JAK1, JAK2 and JAK3.

GLPG0634 analog (Compoun 176) is a broad spectrum JAK inhibitor with IC50 values of <100 nM against JAK1, JAK2 and JAK3.

Specifications

Chemical Name or Material GLPG0634 analog
CAS 1206101-20-3
Color Khaki
Purity Grade Notes Research
Molecular Formula C23H18N6O2
Quantity 50 mg
Solubility Information DMSO : ≥ 53 mg/mL (129.13 mM)
SMILES O=C(C1CC1)NC2=NN3C(C4=CC=C(OCC5=CC=C(C#N)N=C5)C=C4)=CC=CC3=N2
Molecular Weight (g/mol) 410.43
Formula Weight 410.43
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.