missing translation for 'onlineSavingsMsg'
Learn More

Escitalopram oxalate, MedChemExpress

Product Code. 16492147
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16492147 50 mg 50mg
16466008 10 mg 10mg
16446426 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 16492147

Brand: Medchem Express HY14258A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Escitalopram ((S)-Citalopram) oxalate, the S-enantiomer of racemic Citalopram, is a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM. Escitalopram oxalate has ∼30 fold higher binding affinity than its R(-)-enantiomer and shows selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET). Escitalopram oxalate is an antidepressant for the research of major depression.

Escitalopram ((S)-Citalopram) oxalate, the S-enantiomer of racemic Citalopram, is a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM. Escitalopram oxalate has ∼30 fold higher binding affinity than its R(-)-enantiomer and shows selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET). Escitalopram oxalate is an antidepressant for the research of major depression.

Specifications

Chemical Name or Material Escitalopram oxalate
CAS 219861-08-2
Color White
Purity Grade Notes Research
Molecular Formula C22H23FN2O5
Quantity 50 mg
Synonym (S)-Citalopram oxalate(S)-(+)-Citalopram oxalate
Solubility Information H2O : ≥ 14.29 mg/mL (34.48 mM)
SMILES FC1=CC=C([C@@]2(CCCN(C)C)C(C=CC(C#N)=C3)=C3CO2)C=C1.O=C(O)C(O)=O
Molecular Weight (g/mol) 414.43
Formula Weight 414.43
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.