missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enclomiphene citrate, MedChemExpress
Description
Enclomiphene ((E)-Clomiphene) citrate is a potent and orally active non-steroidal estrogen receptor antagonist, with antioestrogenic property. Enclomiphene citrate can be used for the research of ovarian dysfunction, testosterone deficiency, male hypogonadism and type 2 diabetes.
Specifications
Specifications
| Chemical Name or Material | Enclomiphene citrate |
| CAS | 7599-79-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H36ClNO8 |
| Quantity | 50 mg |
| Synonym | (E)-Clomiphene citratetrans-Clomiphene citrateEnclomifene citrate |
| Solubility Information | DMSO : ≥ 50 mg/mL (83.60 mM) |
| SMILES | Cl/C(C1=CC=CC=C1)=C(C2=CC=C(OCCN(CC)CC)C=C2)\C3=CC=CC=C3.O=C(CC(C(O)=O)(O)CC(O)=O)O.[(E)] |
| Molecular Weight (g/mol) | 598.08 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?