missing translation for 'onlineSavingsMsg'
Learn More

Cenicriviroc Mesylate, MedChemExpress

Product Code. 16442367
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442367

Brand: Medchem Express HY14882A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cenicriviroc Mesylate (TAK-652 Mesylate) is a dual CCR2/CCR5 antagonist, also inhibits both HIV-1 and HIV-2, and displays potent anti-inflammatory and antiinfective activity.

Cenicriviroc Mesylate (TAK-652 Mesylate) is a dual CCR2/CCR5 antagonist, also inhibits both HIV-1 and HIV-2, and displays potent anti-inflammatory and antiinfective activity.

Specifications

Chemical Name or Material Cenicriviroc Mesylate
CAS 497223-28-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C42H56N4O7S2
Quantity 50 mg
Synonym TAK-652 Mesylate TBR-652 Mesylate
Solubility Information DMSO : 100 mg/mL (126.10 mM; Need ultrasonic)
SMILES O=C(/C1=C/C2=CC(C3=CC=C(OCCOCCCC)C=C3)=CC=C2N(CC(C)C)CCC1)NC4=CC=C([S@](CC5=CN=CN5CCC)=O)C=C4.OS(=O)(C)=O
Molecular Weight (g/mol) 793.05
Formula Weight 793.05
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.