missing translation for 'onlineSavingsMsg'
Learn More

Cabergoline, MedChemExpress

Product Code. 16432537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16432537 50 mg 50mg
16489017 5 mg 5mg
16486428 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16432537

Brand: Medchem Express HY15296

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cabergoline is an ergot derived-dopamine D2-like receptor agonist that has high affinity for D2, D3, and 5-HT2B receptors (Ki=0.7, 1.5, and 1.2, respectively).

Cabergoline is an ergot derived-dopamine D2-like receptor agonist that has high affinity for D2, D3, and 5-HT2B receptors (Ki=0.7, 1.5, and 1.2, respectively).

Specifications

Chemical Name or Material Cabergoline
CAS 81409-90-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H37N5O2
Quantity 50 mg
Synonym FCE-21336
Solubility Information DMSO : 250 mg/mL (553.59 mM; Need ultrasonic)
SMILES C=CCN1[C@@]2([H])[C@@](C[C@H](C1)C(N(CCCN(C)C)C(NCC)=O)=O)([H])C3=CC=CC4=C3C(C2)=CN4
Molecular Weight (g/mol) 451.6
Formula Weight 451.6
Percent Purity 97.58%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.