missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brompheniramine maleate, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Brompheniramine maleate is a selective histamine H1 receptor antagonist with a Kd of 6.06 nM. Brompheniramine maleate can block the hERG channels, calcium channels, and sodium channels with IC50s of 0.90 μM, 16.12 μM and 21.26 μM, respectively. Brompheniramine maleate has anticholinergic, antidepressant and anesthetic properties and can be used for allergic rhinitis research.
Specifications
Specifications
| Chemical Name or Material | Brompheniramine maleate |
| CAS | 980-71-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H23BrN2O4 |
| Quantity | 50 mg |
| Synonym | (±)-Brompheniramine maleate |
| Solubility Information | DMSO : 100 mg/mL (229.72 mM; Need ultrasonic) ∣H2O : 100 mg/mL (229.72 mM; Need ultrasonic) |
| SMILES | O=C(O)/C=C\C(O)=O.CN(C)CCC(C1=CC=CC=N1)C2=CC=C(Br)C=C2 |
| Molecular Weight (g/mol) | 435.31 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction