missing translation for 'onlineSavingsMsg'
Learn More

BIA 10-2474, MedChemExpress

Product Code. 16443827
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443827 50 mg 50mg
16467878 10 mg 10mg
16417696 100 mg 100mg
16400348 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16443827 Supplier Medchem Express Supplier No. HY19740
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BIA 10-2474 is an inhibitor of fatty acid amide hydrolase (FAAH) with IC50 values of 50 to 70mg/kg in various rat brain regions.

BIA 10-2474 is an inhibitor of fatty acid amide hydrolase (FAAH) with IC50 values of 50 to 70mg/kg in various rat brain regions.

Specifications

Chemical Name or Material BIA 10-2474
CAS 1233855-46-3
Color White
Purity Grade Notes Research
Molecular Formula C16H20N4O2
Quantity 50 mg
Solubility Information DMSO : 2.2 mg/mL (7.32 mM; Need ultrasonic)
SMILES O=C(N1C=C(C2=C[N+]([O-])=CC=C2)N=C1)N(C3CCCCC3)C
Molecular Weight (g/mol) 300.36
Formula Weight 300.36
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.