missing translation for 'onlineSavingsMsg'
Learn More

BAY 73-6691 racemate, MedChemExpress

Product Code. 16480287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480287 50 mg 50mg
16486647 5 mg 5mg
16464956 100 mg 100mg
16403938 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480287 Supplier Medchem Express Supplier No. HY104028A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress BAY 73-6691 racemate is a phosphodiesterase 9 inhibitor extracted from patent WO 2017070293 A1.

BAY 73-6691 racemate is a phosphodiesterase 9 inhibitor extracted from patent WO 2017070293 A1.

Specifications

Chemical Name or Material BAY 73-6691 racemate
Physical Form Solid
CAS 794568-90-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H12ClF3N4O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 160 mg/mL (448.52 mM; Need ultrasonic and warming)
SMILES O=C1C2=C(N(C3=CC=CC=C3Cl)N=C2)NC(CC(C)C(F)(F)F)=N1
Molecular Weight (g/mol) 356.73
Formula Weight 356.73
Percent Purity 99.82%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.