missing translation for 'onlineSavingsMsg'
Learn More

BAY-678, MedChemExpress

Product Code. 16430777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430777 50 mg 50mg
16494458 10 mg 10mg
16463096 25 mg 25mg
16437187 5 mg 5mg
16415346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16430777 Supplier Medchem Express Supplier No. HY111457A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).

BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).

Specifications

Chemical Name or Material BAY-678
Physical Form Solid
CAS 675103-36-3
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H15F3N4O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (249.78 mM) ∣Ethanol : ≥ 4.76 mg/mL (11.89 mM)
SMILES N#CC1=NC=C([C@H](C(C(C)=O)=C(C)N2C3=CC=CC(C(F)(F)F)=C3)NC2=O)C=C1
Molecular Weight (g/mol) 400.35
Formula Weight 400.35
Percent Purity 99.24%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.