missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyperone, MedChemExpress

Product Code. 16434877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16434877 50 mg 50mg
16431488 5 mg 5mg
16423896 25 mg 25mg
16409078 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16434877 Supplier Medchem Express Supplier No. HYN0710
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress alpha-Cyperone (α-Cyperone) is associated with the down-regulation of COX-2, IL-6, Nck-2, Cdc42 and Rac1, resulting in reduction of inflammation, which would be highly beneficial for treatment of inflammatory diseases such as AD.

alpha-Cyperone (α-Cyperone) is associated with the down-regulation of COX-2, IL-6, Nck-2, Cdc42 and Rac1, resulting in reduction of inflammation, which would be highly beneficial for treatment of inflammatory diseases such as AD.

Specifications

Chemical Name or Material alpha-Cyperone
CAS 473-08-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C15H22O
Quantity 50 mg
Synonym α-Cyperone (+)-α-Cyperone
Solubility Information Ethanol : 140 mg/mL (641.23 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (458.02 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1C(C)=C2C[C@H](C(C)=C)CC[C@@]2(C)CC1
Molecular Weight (g/mol) 218.33
Formula Weight 218.33
Percent Purity 98.41%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.