missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Pb/Sn/Sb Babbitt, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Pb/Sn/Sb Babbitt, 50g
Specifications
Specifications
| Chemical Name or Material | Nickel/Iron/Tellurium/Zinc/Indium/Silver/Cadmium/Bismuth/Copper/Arsenic/Tin/Antimony |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Nickel (0.0005) %/Iron (0.001) %/Tellurium (0.008) %/Zinc (0.016) %/Indium (0.021) %/Silver (0.028) %/Cadmium (0.057) %/Bismuth (0.115) %/Copper (0.12) %/Arsenic (0.22) %/Tin (10.4) %/Antimony (16.2) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction