Learn More
Chem-Lab 500ML VANADIUM SOLUTION 1000 PPM
Brand:  Chem-Lab CL01.2208.0500
500ML VANADIUM SOLUTION 1000 PPM
Product Code. 11959142
  • 379.00 NOK / 500mL
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Vanadium standard solution 1000 µg/ml
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric acid