missing translation for 'onlineSavingsMsg'
Learn More

ASTM D7220, Sulfur Set A in Light Oil (20 cSt): S @ 0, 5, 10, 25, 50 μg/g, VHG™

Product Code. 17942026
Change view
Click to view available options
Quantity:
5 x 100 mL
Unit Size:
500mL
Product Code. Quantity unitSize
17942026 5 x 100 mL 500mL
1 options
This item is not returnable. View return policy

Product Code. 17942026

Brand: VHG Labs VHGKTS20MIN7220A5

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ASTM D7220, Sulfur Set A in Light Oil (20 cSt): S @ 0, 5, 10, 25, 50 μg/g
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur
CAS 7704-34-9
Methods XRF/UVF
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/S
SMILES [S]
IUPAC Name sulfur
Solvent or Matrix 20 cSt Oil
Molecular Weight (g/mol) 32.065
Quantity 5 x 100 mL
Formula Weight 31.972 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.