Learn More
Chem-Lab 500ML Aluminium standard solution (Plasma HIQU)(1 g Al / l 2% HNO3)
Brand:  Chem-Lab CL01.0101.0500
500ML Aluminium standard solution (Plasma HIQU)(1 g Al / l 2% HNO3)
Product Code. 11903024
  • 1380.00 NOK / 500mL
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Aluminium standard solution 1000 µg/ml
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid, Aluminium(III) nitrate