missing translation for 'onlineSavingsMsg'
Learn More

Flumatinib mesylate, MedChemExpress

Product Code. 16491706
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
16491706 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 16491706

Brand: Medchem Express HY13905

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Flumatinib (HHGV678) mesylate is an orally active and selective inhibitor of Bcr-Abl. Flumatinib mesylate inhibits c-Abl, PDGFRβ and c-Kit with IC50 values of 1.2, 307.6 and 665.5 nM, respectively. Flumatinib mesylate inhibits Bcr-Abl autophosphorylation and Stat5 and Erk1/2 phosphorylation. Flumatinib mesylate inhibits tumor growth in chronic myelogenous leukemia model.

Flumatinib (HHGV678) mesylate is an orally active and selective inhibitor of Bcr-Abl. Flumatinib mesylate inhibits c-Abl, PDGFRβ and c-Kit with IC50 values of 1.2, 307.6 and 665.5 nM, respectively. Flumatinib mesylate inhibits Bcr-Abl autophosphorylation and Stat5 and Erk1/2 phosphorylation. Flumatinib mesylate inhibits tumor growth in chronic myelogenous leukemia model.

Specifications

Chemical Name or Material Flumatinib mesylate
CAS 895519-91-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H33F3N8O4S
Quantity 500 mg
Synonym HHGV678 mesylate
Solubility Information H2O : 50 mg/mL (75.91 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (75.91 mM; Need ultrasonic)
SMILES O=C(NC1=CC(NC2=NC=CC(C3=CC=CN=C3)=N2)=C(C)N=C1)C4=CC=C(CN5CCN(C)CC5)C(C(F)(F)F)=C4.CS(=O)(O)=O
Molecular Weight (g/mol) 658.69
Formula Weight 658.69
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.