missing translation for 'onlineSavingsMsg'
Learn More

Thalidomide-O-amido-C3-COOH, MedChemExpress

Product Code. 16411576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411576 500 mg 500mg
16481019 1 g 1g
16464916 100 mg 100mg
16430106 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16411576 Supplier Medchem Express Supplier No. HY103612
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Thalidomide-O-amido-C3-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.

Thalidomide-O-amido-C3-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.

Specifications

Chemical Name or Material Thalidomide-O-amido-C3-COOH
CAS 2308035-51-8
Color White
Purity Grade Notes Research
Molecular Formula C19H19N3O8
Quantity 500 mg
Synonym Cereblon Ligand-Linker Conjugates 7 E3 ligase Ligand-Linker Conjugates 15
Solubility Information DMSO : ≥ 55.5 mg/mL (132.98 mM)
SMILES O=C(N(C(CCC1=O)C(N1)=O)C2=O)C(C2=CC=C3)=C3OCC(NCCCC(O)=O)=O
Molecular Weight (g/mol) 417.37
Formula Weight 417.37
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.