missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AICAR, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Description
AICAR (Acadesine) is an adenosine analog and a AMPK activator. AICAR regulates the glucose and lipid metabolism, and inhibits proinflammatory cytokines and iNOS production. AICAR is also an autophagy, YAP and mitophagy inhibitor.
Specifications
Specifications
| Chemical Name or Material | AICAR |
| CAS | 2627-69-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C9H14N4O5 |
| Quantity | 500 mg |
| Synonym | Acadesine AICA Riboside |
| Solubility Information | DMSO : 125 mg/mL (484.06 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 65 mg/mL (251.71 mM; Need ultrasonic and warming) |
| SMILES | OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C=NC(C(N)=O)=C2N)O1 |
| Molecular Weight (g/mol) | 258.23 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction