Learn More
50 NTU Non-Ratio Turbidity Standard, Reagecon
Shop All Reagecon Products
Click to view available options
Volume (Metric):
100 mL
Unit Size:
100mL
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Specifications
| For Use With (Equipment) | Non Ratio Instruments |
| Description | HDPE Bottle |
| Accuracy | ± 1% NTU |
| Turbidity | 50 NTU ± 1% |
| Volume (Metric) | 100 mL |