Learn More
Hamilton™ 50 mL 1000 Series Gas Tight Syringe
Product Code. 17804332
Click to view available options
Needle Gauge:
22 G
No Needle Included
Needle Point Style:
No Needle Included
Sharp, Bevel, Curved, Non-coring
Connection Type:
Luer Lock
Male Luer Lock
Sample Lock
Unit Size:
Each
Product Code. 17804332
Brand: Hamilton™ CAL80008
The 1000 Series syringes offer a mid-volume Gas Tight solution.
  • This syringe series boasts a variety of termination and needle options, providing exceptional versatility and adaptability.
  • Dispenses volumes up to 50 mL.
Specifications
Autoclavable Not Autoclavable
Material Borosilicate Glass
Needle Gauge 22 G
Diameter (English) Inner 1.28 in.
Diameter (English) Inner Needle 0.0161 in.
Diameter (Metric) Inner Needle 0.41 mm
Diameter (English) Outer Needle 0.0283 in.
Diameter (English) Outer 1.45 in.
Diameter (Metric) Inner 32.6 mm
Diameter (Metric) Outer 36.9 mm
Diameter (Metric) Outer Needle 0.72 mm
Length (English) Needle 2 in.
Max. Temperature (English) 176°F
Max. Temperature (Metric) 80°C
Min. Temperature (English) 50°F
Model 1050 SL Syringe
Min. Temperature (Metric) 10°C
Quantity 1
Volume (Metric) 50 mL
Length (Metric) Needle 51 mm
Needle Point Style Sharp, Bevel, Curved, Non-coring
Connection Type Sample Lock