missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Phosphorylribose 1-pyrophosphate sodium salt hydrate, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
Specifications
Specifications
| Color | White |
| Assay Percent Range | 0.75 |
| Quantity | 100 mg |
| Formula Weight | 390.1 |
| Physical Form | Powder |
| Chemical Name or Material | 5-Phosphorylribose 1-pyrophosphate sodium salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction