missing translation for 'onlineSavingsMsg'
Learn More

5-Phosphorylribose 1-pyrophosphate sodium salt hydrate, MP Biomedicals™

Product Code. 11338559 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
This item is not returnable. View return policy

Product Code. 11338559

Brand: MP Biomedicals 0210098125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 108321-05-7
Molecular Formula C5H13NaO14P3
Molecular Weight (g/mol) 413.056
InChI Key CFIMWVBJOCHTCA-UHFFFAOYSA-N
Synonym alpha-d-ribosyldiphosphate5-phosphatepentasodiumsalt
PubChem CID 133611921
IUPAC Name [3,4-dihydroxy-5-(phosphonooxymethyl)oxolan-2-yl] phosphono hydrogen phosphate;sodium
SMILES C(C1C(C(C(O1)OP(=O)(O)OP(=O)(O)O)O)O)OP(=O)(O)O.[Na]

Specifications

Color White
Assay Percent Range 0.75
Quantity 25 mg
Formula Weight 390.1
Physical Form Powder
Chemical Name or Material 5-Phosphorylribose 1-pyrophosphate sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.