Learn More
5-(Phenylmethoxy)-1H-indole-3-ethan-Alpha,Alpha,Beta,Beta-d4-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(Phenylmethoxy)-1H-indole-3-ethan-alpha,alpha,beta,beta-d4-ol |
| CAS | 75238-47-0 |
| Molecular Formula | C17 D4 H13 N O2 |
| Quantity | 5 mg |
| Synonym | 1H-Indole-3-ethan-α,α,β,β-d4-ol','5-(phenylmethoxy)- (9CI), 5-(Phenylmethoxy)-1H-indole-3-ethan-α,α,β,β-d4-ol, 2-(5-Benzyloxy-1H-indol-3-yl)-ethanol, 5-Benzyloxytryptophol-α,α,β,β-d4, 5-Benzyloxy-1H-indole-3-ethanol-D4 (ethanol-D4) |
| InChI Formula | InChI=1S/C17H17NO2/c19-9-8-14-11-18-17-7-6-15(10-16(14)17)20-12-13-4-2-1-3-5-13/h1-7,10-11,18-19H,8-9,12H2/i8D2,9D2 |
| SMILES | [2H]C([2H])(O)C([2H])([2H])c1c[nH]c2ccc(OCc3ccccc3)cc12 |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(5-phenylmethoxy-1H-indol-3-yl)ethanol |
| Molecular Weight (g/mol) | 271.347 |
| Formula Weight | 271.151 |