Learn More
5-Nitroso-2,4,6-triaminopyrimidine, TRC
Product Code. 30464552
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464552 1 g 1g
30472308 5 g -
30479422 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30464552 Supplier Toronto Research Chemicals Supplier No. TRCN5469751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1006-23-1
Molecular Formula C4 H6 N6 O
Molecular Weight (g/mol) 154.13
Synonym 5-Nitrosopyrimidine-2,4,6-triamine, Triamterene Imp. A (EP), Nitrosotriaminopyrimidine
IUPAC Name 5-nitrosopyrimidine-2,4,6-triamine
SMILES Nc1nc(N)c(N=O)c(N)n1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C4H6N6O/c5-2-1(10-11)3(6)9-4(7)8-2/h(H6,5,6,7,8,9)
Formula Weight 154.0603
Chemical Name or Material Nitrosotriaminopyrimidine