Learn More
5-Nitro-8-hydroxyquinoline, TRC
Product Code. 30040371
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040371 10 g 10g
18060020 100 g 100g
17959829 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30040371 Supplier Toronto Research Chemicals Supplier No. TRCN49621510G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4008-48-4
Molecular Formula C9 H6 N2 O3
Molecular Weight (g/mol) 190.16
Synonym 8-Quinolinol, 5-nitro-, 5-(Hydroxyl[oxido]amino)-8-hydroxyquinoline, 5-Nitro-8-hydroxyquinoline, 5-Nitro-8-oxychinoline, 5-Nitro-8-oxyquinoline, 5-Nitro-8-quinolinol, 5-Nitrox, 5-Nitroxine, 5NOK, 8-Hydroxy-5-nitroquinoline, A 82, BAS 58, NSC 74947, NSC 74948, Nibiol, Nitroxolin, Nitroxoline, Noxibiol
IUPAC Name 5-nitroquinolin-8-ol
SMILES Oc1ccc(c2cccnc12)[N+](=O)[O-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C9H6N2O3/c12-8-4-3-7(11(13)14)6-2-1-5-10-9(6)8/h1-5,12H
Formula Weight 190.0378
Chemical Name or Material Nitroxoline