Learn More
5-Nitro-1,10-phenanthroline, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4199-88-6 |
| Molecular Formula | C12H7N3O2 |
| MDL Number | MFCD00004981 |
| Quantity | 1 g |
| SMILES | [O-][N+](=O)C1=CC2=CC=CN=C2C2=C1C=CC=N2 |
| Molecular Weight (g/mol) | 225.20 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-Nitro-1,10-phenanthroline |