Learn More
5'-N-Ethylcarboxamidoadenosine, MedChemExpress
Product Code. 16493798
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16493798
Brand: Medchem Express HY103173
MedChemExpress 5'-N-Ethylcarboxamidoadenosine (NECA) is a nonselective adenosine receptor agonist.
5'-N-Ethylcarboxamidoadenosine (NECA) is a nonselective adenosine receptor agonist.
Specifications
Chemical Name or Material 5'-N-Ethylcarboxamidoadenosine
CAS 35920-39-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H16N6O4
Quantity 10 mg
Synonym NECA
Solubility Information DMSO : 62.5 mg/mL (202.73 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O[C@H]1[C@H](N2C=NC3=C(N)N=CN=C23)O[C@H](C(NCC)=O)[C@H]1O
Molecular Weight (g/mol) 308.29
Formula Weight 308.29
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.