Learn More
5-Methyltryptamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Methyltryptamine Hydrochloride |
| CAS | 1010-95-3 |
| Molecular Formula | C11H15ClN2 |
| Quantity | 2 g |
| Synonym | 5-Methyl-1H-Indole-3-ethanamine Hydrochloride, 3-(2-Aminoethyl)-5-methylindole Hydrochloride |
| InChI Formula | InChI=1S/C11H14N2.ClH/c1-8-2-3-11-10(6-8)9(4-5-12)7-13-11;/h2-3,6-7,13H,4-5,12H2,1H3;1H |
| SMILES | Cl.Cc1ccc2[nH]cc(CCN)c2c1 |
| IUPAC Name | 2-(5-methyl-1H-indol-3-yl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 210.7 |
| Formula Weight | 210.0924 |