Learn More
5-Methylpyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Methylpyrimidine |
| CAS | 2036-41-1 |
| Molecular Formula | C5 H6 N2 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C5H6N2/c1-5-2-6-4-7-3-5/h2-4H,1H3 |
| SMILES | Cc1cncnc1 |
| IUPAC Name | 5-methylpyrimidine |
| Molecular Weight (g/mol) | 94.11 |
| Formula Weight | 94.0531 |