missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylbenzimidazole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.
Solubility
Soluble in chloroform (25mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.
Solubility
Soluble in chloroform (25mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 112°C to 117°C |
Boiling Point | 169°C to 172°C (1 mmHg) |
Quantity | 5 g |
Beilstein | 2628 |
Solubility Information | Soluble in chloroform (25mg/ml). |
Formula Weight | 132.17 |
Percent Purity | 98% |
Chemical Name or Material | 5-Methylbenzimidazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'