Learn More
5-Methyl-sorbic Acid Ethyl Ester, TRC
Product Code. 30474696
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474696 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30474696 Supplier Toronto Research Chemicals Supplier No. TRCM326840100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methyl-sorbic Acid Ethyl Ester
CAS 10231-96-6
Molecular Formula C9H14O2
Quantity 100 mg
Synonym Ethyl 5-Methyl-2,4-hexadienoate, 5-Methyl-2,4-hexadienoic Acid Ethyl Ester
InChI Formula InChI=1S/C9H14O2/c1-4-11-9(10)7-5-6-8(2)3/h5-7H,4H2,1-3H3/b7-5+
SMILES CCOC(=O)\C=C\C=C(C)C
IUPAC Name ethyl (2E)-5-methylhexa-2,4-dienoate
Molecular Weight (g/mol) 154.21
Formula Weight 154.0994