Learn More
5-Methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 849776-88-1 |
| Molecular Formula | C16H21BN2O2 |
| MDL Number | MFCD06797463 |
| Quantity | 1 g |
| SMILES | CC1(C)C(C)(C)OB(C2=C(C)N(C3=CC=CC=C3)N=C2)O1 |
| Molecular Weight (g/mol) | 284.16 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-Methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole |