Learn More
5 Lipoxygenase Mouse anti-Human, Porcine, Clone: 3B1B7, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | 5 Lipoxygenase |
|---|---|
| Clone | 3B1B7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16898095
|
Proteintech
66326-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16888095
|
Proteintech
66326-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The enzyme 5-lipoxygenase (5-LO) plays a key role in regulating the production of leukotrienes (LT) (Funk, 2001). Overproduction of LT contributes to several diseases, most notably chronic inflammatory diseases, including asthma (Drazen et al., 1994), fibrosis (Wilborn et al., 1996) and atherosclerosis (Dwyer et al., 2004). Recent work has demonstrated that the activity of 5-LO is regulated by PKA phosphorylation of serine-523 in 5-LO (Luo et al., 2004). Under normal conditions, this phosphorylation may be important in limiting inflammation. Abnormal signaling through cAMP and PKA, then, could contribute to a variety of diseases, including those characterized by chronic inflammation. The phospho-specific antibody to Ser523 on 5-LO is thus likely to provide a valuable toll for studies of the role of 5-LO regulation in diseases such as asthma, fibrosis and atherosclerosis.Specifications
| 5 Lipoxygenase | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 5 lipoxygenase, 5 LO, 5 LOX, 5LPG, ALOX5, arachidonate 5 lipoxygenase, LOG5 | |
| ALOX5 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 3B1B7 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Pig, Human | |
| P09917 | |
| 100156205, 240 | |
| 5 Lipoxygenase Fusion Protein Ag18168 | |
| Primary | |
| -20°C | |
| ALOX5 |