Learn More
(5-Isopropyl-2-methoxyphenyl)boronic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 216393-63-4 |
| Molecular Formula | C10H15BO3 |
| MDL Number | MFCD01318154 |
| Quantity | 1 g |
| SMILES | COC1=C(C=C(C=C1)C(C)C)B(O)O |
| Molecular Weight (g/mol) | 194.04 |
| Percent Purity | 98% |
| Chemical Name or Material | (5-Isopropyl-2-methoxyphenyl)boronic acid |